Learn More
Hach Lange™ Cella in vetro per campioni
Codice prodotto. 15173852
Cambia vista
Click to view available options
Da utilizzare con (apparecchiatura):
2100 N, 2100 AN e turbidimetri da laboratorio RATIO
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Da utilizzare con (apparecchiatura) Dimensione della confezione
15173852 2100 N, 2100 AN e turbidimetri da laboratorio RATIO Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15173852 Fornitore Hach Lange™ N. del fornitore 2084900
Spedizione stimata: 30-09-2026
Easy to use and cost effective replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters are made of glass and contain six sample cell vials with caps.
  • For use with Hach Ratio 18900, 2100N and 2100AN Laboratory Turbidmeters
  • Replacement sample cells for use with 2100N, 2100AN, and lab Ratio turbidimeters
  • For use with Hach Ratio 18900, 2100N and 2100AN laboratory turbidmeters
  • Includes six sample cell vials with caps
  • Made of glass material\
TRUSTED_SUSTAINABILITY
Specifica
Tipo Cella in vetro per campioni
Da utilizzare con (apparecchiatura) 2100 N, 2100 AN e turbidimetri da laboratorio RATIO