Learn More
Hecht Karl™ Glass Shot
Product Code. 10368911
Change view
Click to view available options
Diameter (Metric):
8 mm
10 mm
Unit Size:
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
10368911 10 mm 1kg
10777322 8 mm 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10368911 Supplier Hecht Karl™ Supplier No. 41401010
  • ONLINE EXCLUSIVE PRICE
    40.26€
    35.48€ / 1kg
    Risparmia 4.78€
    11% Sconto
Spedizione stimata: 17-12-2026