Learn More
Kova Plastics by Alltrista Glasstic™ Slide 10 with Grids
Product Code. 15292826
Encompass_Preferred
Change view
Click to view available options
Quantity:
100
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15292826 100 Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15292826 Supplier Kova Plastics by Alltrista Supplier No. 87144E
 Restricted Item
Spedizione stimata: 22-12-2026
A precision manufactured, 10-chambered disposable microscope slide made of optically clear plastic with a Neubauer counting grid.
  • The slide grid is in the focal plane of the specimen, allowing for use over a range of magnifications
  • Reduces pipetting errors and eliminates variables that can interfere with microscopic urinalysis
  • Superior optical plastic with a refractive index that is similar to glass
  • Manufactured to the specifications of high quality optical lenses, smooth with a tolerance below 50
  • Perimeter contained wells for precise volume control and protection against cross-contamination and leakage
  • Easier and faster counting with Neubauer-style ruling pattern
  • Capillary action provides reproducible precision
  • Homogeneous suspension of sample sediments
Specifications
Frosted Not Frosted
Material Optical Plastic
Type Slide with Counting Grid
Well Count 10
Color Transparent
Description Neubauer style ruling pattern
For Use With (Application) Microscopy applications
Quantity 100