Learn More
Glassy carbon spherical powder, 630 to 1000μm, type 1, Alfa Aesar™
Used as such in purifying metals, in electrodes, electrical devices & steel. Glassy Carbon Spherical Powder has numerous applications due its fundamental softness, including in lubricant formulations and as an additive to structural materials and metallic alloys.
Marca: Alfa Aesar 041497.18
Dettagli aggiuntivi : CAS : 7440-44-0 Peso : 0.05000kg Transport : UN number : 1325 Chem class : 4.1 Pack group : III
Quantità | 50g |
---|
Identificatori chimici
MFCD00133992 |
Specifica
Glassy carbon spherical powder | |
50g | |
UN1325 |
Plastic bottle | |
MFCD00133992 | |
Insoluble in water. |