Learn More
Sanplatec™ Glove Box Mobile Lab Space, MBLS-2
Codice prodotto. 30242879
Cambia vista
Click to view available options
Materiale:
PVC
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Materiale Dimensione della confezione
30242879 PVC Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
  • The gloves and replacement gloves provided are simple gloves.For gas replacement, please use gas replacement gloves (Part No. 00193).
Codice prodotto. 30242879 Fornitore Sanplatec™ N. del fornitore 27001
  • ONLINE EXCLUSIVE PRICE
    359.00€
    335.28€ / pezzi 1
    Risparmia 23.72€
    6% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 08-02-2027
Aggiungi al carrello
Aggiungi al carrello
  • The gloves and replacement gloves provided are simple gloves.For gas replacement, please use gas replacement gloves (Part No. 00193).
Lightweight, easy-to-carry foldable glove box. Fully open front for convenient loading and unloading. Simple gas displacement is also possible.
  • The front opening is wide, making it easy to take things in and out.
  • It is easy to take on and off gloves.
  • Lightweight and foldable for easy carrying.
  • Foldable for storage.
  • It can also be used as a simple gas purge.
  • The supplied glove and replacement glove are simple disposable gloves.
  • Please use the Gas Purge glove (code 00193) for Gas Purge.
Specifica
Product Type Glove Box
Tipo di filtro None
Tipo di illuminazione None
Materiale PVC
Peso (sistema metrico) 1.7 kg