missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GlowQuant™ ATP Cell Viability Assay Kit

Codice prodotto. 31934709
Cambia vista
Click to view available options
Quantità:
10 mL
100 mL
Dimensione della confezione:
100mL
10mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
31934709 10 mL 10mL
31934241 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 31934709 Fornitore Invitrogen™ N. del fornitore ATPCV1010
 Articolo soggetto a restrizioni
Spedizione stimata: 31-12-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The GlowQuant™ ATP Cell Viability Assay Kit is a ready-to-use, bioluminescence-based solution for rapid and sensitive measurement of viable cells through ATP detection and quantification.

The GlowQuant™ ATP Cell Viability Assay Kit is a ready-to-use, bioluminescence-based solution for rapid and sensitive measurement of viable cells through ATP detection and quantification. Designed for wide variety of applications, this luminescence-based ATP assay supports cell health, proliferation studies, and cytotoxicity analysis with a simple one-step workflow.

The assay uses firefly luciferase to catalyze the oxidation of luciferin in the presence of ATP and oxygen, generating a stable bioluminescent signal directly proportional to ATP concentration, and therefore to the number of viable cells.

By measuring intracellular ATP (a universal marker of metabolically active cells), this assay provides a direct and quantitative assessment of cell health.

Key Features:

Features of GlowQuant™ ATP Cell Viability Assay Kit include:

  • High sensitivity ATP detection— Detects low cell numbers, down to ~50 cells/well in a 96-well plate.
  • Fast and simple — One-step add-and-read format with minimal hands-on time and results in as little as 10 minutes
  • No reagent preparation required — Supplied as a ready-to-use, pre-formulated reagent.
  • Stable signal — Bioluminescent glow with a signal half-life > 2 hours for flexible plate reading.
  • Quantitative and reliable performance — Linear response over a wide dynamic range for reliable cell-number estimation.
  • Microplate reader compatible — Designed for use with standard luminescence microplate readers in 96- and 384-well formats; compatible with automation.

How it Works

The GlowQuant™ assay features a simple add-and-read protocol with no separate lysis step required.

  1. Add an equal volume of reagent directly to cultured cells.
  2. Mix for 1 minute.
  3. Incubate at room temperature for 10–20 minutes.
  4. Measure luminescence using a luminometer or compatible microplate reader.

The emitted luminescent signal is proportional to intracellular ATP levels. Because viable cells maintain ATP while damaged or non-viable cells rapidly lose ATP, signal intensity directly reflects the number of metabolically active cells in each well.

Supporting Your Cell Health Research Across Detection Modes

Thermo Fisher Scientific supports cell health research with a broad portfolio of validated assays spanning luminescence, fluorescence, and absorbance detection modes. From rapid ATP-based viability assays to multiplex-compatible fluorescent and colorimetric solutions, our cell health tools are designed to integrate seamlessly with standard plate readers, automated workflows, and high-throughput screening platforms— to help provide scalable solutions backed by global technical support.

TRUSTED_SUSTAINABILITY

Specifica

Tipo di cellula Adherent and Suspension Cells
Content And Storage Store at -20°C, protected from light
Metodo di rilevamento Luminescence
Da utilizzare con (applicazione) Cell Viability
Da utilizzare con (apparecchiatura) Luminometer, Microplate Reader
Product Type Cell Viability Reagent
Forma Liquid
Formato 96-well Plate, 384-well Plate
Tempo di incubazione 10 to 20 min.
Product Line GlowQuant
Quantità 10 mL
Research Category Cell Health
Vedi altri risultati Mostra meno risultati

For Research Use Only.

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.