Learn More
Abcam Glucose-6-Phosphate Assay Kit - High Sensitivity (Fluorometric)
Product Code. 32116718 Shop All Abcam Products
Change view
Click to view available options
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
32116718 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32116718 Supplier Abcam Supplier No. ab107923100Test
 Restricted Item
Spedizione stimata: 25-11-2026
Measure 6-O-phosphono-alpha-D-glucopyranose in 20m in Biofluids, Cell/Tissue Extracts, Cell Culture Media with Glucose-6-Phosphate Assay Kit AB107923. Sensitivity>1 μM. Cited in 3 publications. Use with Microplate reader.
Measure 6-O-phosphono-alpha-D-glucopyranose in 20m in Biofluids, Cell/Tissue Extracts, Cell Culture Media with Glucose-6-Phosphate Assay Kit AB107923. Sensitivity>1 μM. Cited in 3 publications. Use with Microplate reader.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantity 100 Tests
Regulatory Status RUO