missing translation for 'onlineSavingsMsg'
Learn More

Glut1 Antibody (rGLUT1/2476), HRP, R&D Systems™

Product Code. 18608117 Shop All Bio Techne Products
Change view
Click to view available options
Quantità:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
18608117 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18608117 Supplier Novus Biologicals Supplier No. NBP308479H
 Articolo soggetto a restrizioni
Spedizione stimata: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

Glut1 Monoclonal specifically detects Glut1 in Human samples. It is validated for ELISA/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigene Glut1
Applicazioni ELISA, Immunohistochemistry (Paraffin)
Classificazione Monoclonal
Clone rGLUT1/2476
Coniugato HRP
Formulazione PBS
Alias gene Choreoathetosis/Spasticity, Episodic (Paroxysmal Choreoathetosis), CSE, DYT17, DYT18, DYT9, EIG12, Glucose transporter type 1, erythrocyte/brain, GLUT-1, GLUT1DS, HepG2 glucose transporter, HTLVR, Human T-Cell Leukemia Virus (I and II) Receptor, MGC141895, MGC141896, PED, solute carrier family 2 (facilitated glucose transporter), member 1, Solute Carrier Family 2 Member 1, solute carrier family 2, facilitated glucose transporter member 1
Specie ospite Mouse
Immunogeno Recombinant fragment of human Glut1 protein (around aa 203-305) (exact sequence is proprietary) (Uniprot: P11166)
Metodo di purificazione Protein A or G purified
Quantità 100 μg
Status giuridico RUO
Disciplina di ricerca Cancer, Cardiovascular Biology, Cellular Markers, Diabetes Research, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroscience, Plasma Membrane Markers, Signal Transduction
Primario o secondario Primary
ID gene (immissione) 6513
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Forma Purified
Isotype IgG1 κ
Linea di prodotti Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.