Learn More
Novus Biologicals™ Glutathione Peroxidase 1/GPX1 Protein
Codice prodotto. 18597552 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
0.5 mg
Dimensione della confezione:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18597552 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18597552 Fornitore Novus Biologicals™ N. del fornitore NBP1725120.5mg
Please to purchase this item. Need a web account? Register with us today!
Highly purified. Generating reliable and reproducible results.
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-203 of Human Glutathione Peroxidase 1/GPX1 The Recombinant Human Glutathione Peroxidase 1/GPX1 Protein is derived from E. coli. The Recombinant Human Glutathione Peroxidase 1/GPX1 Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY
Specifica
Da utilizzare con (applicazione) SDS-PAGE
Formulazione Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 2mM DTT, 30% glycerol, 100mM NaCl
Peso molecolare 24.2kDa
Metodo di purificazione SDS-PAGE
Quantità 0.5 mg
Sorgente Human
Requisiti di stoccaggio Store at -80°C. Avoid freeze-thaw cycles.
Status giuridico RUO
Nome comune Glutathione Peroxidase 1/GPX1
Coniugato Unconjugated
Purity or Quality Grade >90%