missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Gly-Arg-Gly-Asp-Ser

Codice prodotto. 16414036
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
25 mg
Dimensione della confezione:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16414036 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 16414036 Fornitore MedChemExpress N. del fornitore HYP0295

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

MedChemExpress Gly-Arg-Gly-Asp-Ser is a pentapeptide that forms the cell-binding domain of a glycoprotein, osteopontin. Gly-Arg-Gly-Asp-Ser binds to integrin receptors αvβ3 and αvβ5 with estimated IC50 of ∼5 and ∼6.5 μM

Specifications

Purezza 95.05%
CAS 96426-21-0
Content And Storage Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Peso molecolare 490.47 g/mol
Product Type Peptides
Quantità 25 mg
Shipping Condition Room temperature
Solubilità DMSO : 50 mg/mL (101.94 mM∣Need ultrasonic) ∣H2O : 33.33 mg/mL (67.96 mM∣Need ultrasonic)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.