Learn More
Glycerol (Ultra Pure 99.5%)
Product Code. 11401572 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11401572 500 mL 500mL
11411572 1 L 1 L
11421572 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11401572 Supplier MP Biomedicals Supplier No. 04800687
  • ONLINE EXCLUSIVE PRICE
    111.00€
    105.00€ / 500mL
    Risparmia 6.00€
    5% Sconto
View all promotions
 Restricted Item
In stock
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Chemical Identifiers
CAS 56-81-5
Molecular Formula C3H8O3
Molecular Weight (g/mol) 92.09
MDL Number MFCD00004722
InChI Key PEDCQBHIVMGVHV-UHFFFAOYSA-N
Synonym glycerol, glycerin, glycerine, 1,2,3-propanetriol, glycyl alcohol, trihydroxypropane, glyceritol, propanetriol, 1,2,3-trihydroxypropane, osmoglyn
PubChem CID 753
ChEBI CHEBI:17754
IUPAC Name propane-1,2,3-triol
SMILES OCC(O)CO
Specifications
Quantity 500 mL
Grade Ultrapure
Physical Form Liquid
Chemical Name or Material Glycerol (Ultra Pure 99.5%)