Learn More
Glycine Benzyl Ester-13C2,15N p-Toluenesulfonate, TRC
Specifica
Specifica
| Materiale o nome chimico | Glycine Benzyl Ester-13C2,15N p-Toluenesulfonate |
| CAS | 874462-68-7 |
| Formula molecolare | C1413C2H1915NO5S |
| Quantità | 25 mg |
| Sinonimo | Glycine Phenylmethyl Ester, 4-Methylbenzenesulfonate-13C2,15N, Benzyl Glycinate p-Toluenesulfonate-13C2,15N, Glycine Benzyl Ester p-Tosylate-13C2,15N, Benzyl Glycina-13C2,15N |
| InChI Formula | InChI=1S/C9H11NO2.C7H8O3S/c10-6-9(11)12-7-8-4-2-1-3-5-8;1-6-2-4-7(5-3-6)11(8,9)10/h1-5H,6-7,10H2;2-5H,1H3,(H,8,9,10)/i6+1,9+1,10+1; |
| SMILES | Cc1ccc(cc1)S(=O)(=O)O.[15NH2][13CH2][13C](=O)OCc2ccccc2 |
| IUPAC Name | benzyl 2-(^{15}N)azanylacetate;4-methylbenzenesulfonic acid |
| Molecular Weight (g/mol) | 340.37 |
| Peso formulazione | 340.1021 |