missing translation for 'onlineSavingsMsg'
Learn More

GNAL Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA550981

Dettagli aggiuntivi : Peso : 0.01000kg

Codice prodotto. 15634905

  • 472.00€ / 100µL
Spedizione stimata: 18-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

GNAL Polyclonal Antibody detects endogenous levels of GNAL protein.

GNAL Polyclonal antibody specifically detects GNAL in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Western Blot.
Specifica

Specifica

GNAL
Polyclonal
Unconjugated
Gnal
2610011C15Rik; 9630020G10Rik; adenylate cyclase-stimulating G alpha protein, olfactory type; DYT25; G alpha 10; G protein subunit alpha L; Galphaolf; Gna10; Gnal; Golf; guanine nucleotide binding protein (G protein), alpha activating activity polypeptide, olfactory type; guanine nucleotide binding protein (G protein), alpha stimulating activity polypeptide, olfactory type; guanine nucleotide binding protein, alpha 10; guanine nucleotide binding protein, alpha stimulating, olfactory type; guanine nucleotide-binding protein G(olf) subunit alpha; guanine nucleotide-binding protein G(olf), alpha subunit; Olf; olfa; Olf-alpha protein (olfactory neuron-specific G protein); RGD1305940
Rabbit
Antigen affinity chromatography
RUO
14680, 24611, 2774
-20° C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
P38405, P38406, Q8CGK7
Gnal
Synthesized peptide derived from the Internal region of human GA olf
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali