missing translation for 'onlineSavingsMsg'
Learn More

GNB5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA565377

Dettagli aggiuntivi : Peso : 0.01000kg

Codice prodotto. 15908134

  • 514.00€ / 100µL
Spedizione stimata: 17-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: DNKCSVYPLTF DKNENMAAKK KSVAMHTNYL SACSFTNSDM QILTASGDGT Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

GNB5 Polyclonal antibody specifically detects GNB5 in Human samples. It is validated for Immunocytochemistry.
Specifica

Specifica

GNB5
Polyclonal
Unconjugated
Gnb5
flail; flailer; flr; G protein subunit beta 5; G protein, beta subunit 5L; G protein, beta-5 subunit; GB5; GBB5; Gbeta5; GBS; Gnb5; guanine nucleotide binding protein (G protein), beta 5; guanine nucleotide binding protein (G protein), beta polypeptide 5; guanine nucleotide binding protein, beta 5; guanine nucleotide-binding protein subunit beta-5; guanine nucleotide-binding protein, beta subunit 5L; guanine nucleotide-binding protein, beta-5 subunit; hug; transducin beta chain 5
Rabbit
Antigen affinity chromatography
RUO
10681
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O14775
Gnb5
Recombinant Human GNB5
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

For Research Use Only