missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG F(ab')2 Secondary Antibody

Codice prodotto. 11805905
Cambia vista
Click to view available options
Quantità:
2 mg
Dimensione della confezione:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11805905 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11805905 Fornitore Invitrogen™ N. del fornitore 31166

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Goat Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31166 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31166 reacts with the F(ab')2/Fab protion of mouse IgG and it also reacts with the light chains of other mouse immunoglobulins, but does not react against the Fc portion of mouse IgG or non-immunoglobulin serum proteins. This antibody may cross-react with immunoglobulins from other species. Store product at 4°C until opened. After opening, dilute only enough product for a single day's use. Store remainder at 2-8 °C under sterile conditions. Country of Origin: USA.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifica

Antigene Mouse IgG F(ab')2
Applicazioni Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 2.4 mg/mL
Coniugato Unconjugated
Formulazione PBS with no preservative; pH 7.6
Specie ospite Goat
Metodo di purificazione Antigen affinity chromatography
Quantità 2 mg
Status giuridico RUO
Primario o secondario Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.