Learn More
Invitrogen™ Goat anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, DyLight™ 680
Descrizione
Product # 35519 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35519 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. The antibody has been tested by solid-phase adsorbed to ensure minimal cross-reactivity with human, bovine, horse, rabbit, swine, goat, and rat serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 680 Amax= 682 nm; Emax= 715 nm. Mole Dye/Mole Protein Ratio is lot-dependent.
Specifica
Specifica
| Antigene | Mouse IgG (H+L) Cross-Adsorbed |
| Applicazioni | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classificazione | Polyclonal |
| Concentrazione | 1 mg/mL |
| Coniugato | DyLight 680 |
| Formulazione | PBS with 0.02% sodium azide; pH 7.2 |
| Specie ospite | Goat |
| Metodo di purificazione | Antigen affinity chromatography |
| Quantità | 500 μL |
| Status giuridico | RUO |
| Vedi altri risultati |
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.