missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG (H+L) Secondary Antibody, DyLight™ 488

Codice prodotto. 10688674
Cambia vista
Click to view available options
Quantità:
1 mL
Dimensione della confezione:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
10688674 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 10688674 Fornitore Invitrogen™ N. del fornitore 35502
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Edge
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Goat Polyclonal Secondary Antibody

Product # 35502 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 35502 reacts with the heavy chains of mouse IgG and with the light chains common to most mouse immunoglobulins, but does not react against non-immunoglobulin serum proteins. However, this antibody may cross-react with immunoglobulins from other species. Store product protected from light at 4°C until opened. To extend the shelf-life of this product, add an equal volume of glycerol to make a final concentration of approximately 50% glycerol and store at -20°C. DyLight 488 Amax= 493 nm; Emax= 518 nm. Mole Dye/Mole Protein Ratio is lot-dependent.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifica

Antigene Mouse IgG (H+L)
Applicazioni Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato DyLight 488
Formulazione 1 ml di anticorpo come liquido di colore giallo alla concentrazione di 1 mg/ml in tampone fosfato salino 100 mM, pH 7.2, contenente 1% di BSA e 0.02% di sodio azide.
Specie ospite Goat
Immunogeno Purified Mouse IgG, whole molecule.
Metodo di purificazione Antigen affinity chromatography
Quantità 1 mL
Status giuridico RUO
Primario o secondario Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.