missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, FITC, MP Biomedicals
Marca: MP Biomedicals 0855514
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Host Species: Goat
Species Reactivity: Mouse
Specifica
IgG, Whole Molecule | |
FITC | |
2 mg | |
Mouse |
Polyclonal | |
Goat | |
Secondary | |
Purified |