missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Mouse IgG, Whole Molecule Polyclonal Secondary Antibody, HRP, MP Biomedicals
Marca: MP Biomedicals 0855563
Dettagli aggiuntivi : Peso : 0.00200kg
Descrizione
Host Species: Goat
Species Reactivity: Mouse
Specifica
IgG, Whole Molecule | |
HRP | |
Affinity Purified | |
Secondary |
Polyclonal | |
Goat | |
2 mL | |
Mouse |