missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Goat anti-Mouse IgG2c Secondary Antibody

Codice prodotto. 17169645
Cambia vista
Click to view available options
Quantità:
1 mg
Dimensione della confezione:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17169645 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17169645 Fornitore Invitrogen™ N. del fornitore SA510267

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Goat Polyclonal Secondary Antibody

This antibody will react with mouse IgG2c as determined by ELISA and IEP techniques and has less than 1% cross reactivity with mouse IgG1, IgG2b, and IgG3. This antibody may cross-react with IgG from other species.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifica

Antigene Mouse IgG2c
Applicazioni ELISA, Immunoelectrophoresis, Immunohistochemistry, Western Blot
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione PBS with 0.09% sodium azide; pH 7.2
Specie ospite Goat
Immunogeno Mouse IgG2c.
Metodo di purificazione Antigen affinity chromatography
Quantità 1 mg
Status giuridico RUO
Primario o secondario Secondary
Target Species Mouse
Content And Storage 4°C
Product Type Secondary Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.