Learn More
SCHOTT AG Lighting and Imaging™ Gooseneck Light Guide for KL300 LED
Codice prodotto. 11842442 Sfoglia Tutto Schott Prodotti
Cambia vista
Click to view available options
Dimensione della confezione:
Pezzo
Descrizione:
Single Branch Fiber Optic Lightguide
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensione della confezione Descrizione
11842442 Pezzo -
11832442 Pezzo Single Branch Fiber Optic Lightguide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11842442 Fornitore SCHOTT AG Lighting and Imaging™ N. del fornitore 170202
  • ONLINE EXCLUSIVE PRICE
    280.65€
    254.00€ / pezzi 1
    Risparmia 26.65€
    9% Sconto
Spedizione stimata: 29-12-2026
Fiber optic flex and stay light guide in a black lacquered semi-rigid gooseneck tubing.
  • Light guide with eco-friendly Puravis fiber optics.
  • Precise positioning offers complete control of light placement.
  • Gooseneck branches positioned in opposite directions create near shadow-free illumination.
  • Re-adjust with ease.
  • For use with Cold light source KL300LED.
  • Diameter: End tip: 6 mm, Lightsource adapter: 9 mm, Active Fiber optic bundle: 4.5 mm, Outer Jacketing: 6.5 mm
Specifica
Lunghezza (sistema metrico) 500 mm