missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17219-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Mitochondrial glycerophosphate dehydrogenase (EC 1.1.99.5), or GPD2, is located on the outer surface of the inner mitochondrial membrane and catalyzes the unidirectional conversion of glycerol-3-phosphate (G-3-P) to dihydroxyacetone phosphate (DHAP) with concomitant reduction of the enzyme-bound FAD. Together with a cytosolic NAD-linked GPD (GPD1; MIM 138420), GPD2 forms the glycerol phosphate shuttle, which uses the interconversion of G-3-P and DHAP to transfer reducing equivalents into mitochondria, resulting in the reoxidation of NADH formed during glycolysis.Specifica
GPD2 | |
Polyclonal | |
Unconjugated | |
GPD2 | |
GDH2, GPD M, GPD2, GPDH M, GPDM, mGPDH, mtGPD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14571, 25062, 2820 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P35571, P43304, Q64521 | |
Gpd2 | |
GPD2 Fusion Protein Ag11036 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |