missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPRC5C Polyclonal Antibody

Descrizione
Immunogen sequence: TSVYQPTEMA LMHKVPSEGA YDIILPRATA NSQVMGSANS TLRAEDMYSA QSHQAATPPK DGKNSQVFRN Highest antigen sequence indentity to the following orthologs: Mouse - 91%, Rat - 90%.
This polyclonal antibody was raised in Rabbit against GPRC5C. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPRC5C. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Specifica
Specifica
| Antigene | GPRC5C |
| Applicazioni | Immunohistochemistry (Paraffin), Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 0.2 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GPRC5C |
| N. accesso geni | Q9NQ84 |
| Alias gene | 1110028I06Rik; 3200002M13Rik; G protein-coupled receptor class C group 5 member C; G protein-coupled receptor, class C, group 5, member C; G protein-coupled receptor, family C, group 5, member C; Gprc5c; G-protein coupled receptor family C group 5 member C; MGC131820; orphan G-protein coupled receptor; PSEC0087; RAIG3; RAIG-3; retinoic acid inducible protein 3; retinoic acid responsive gene protein; retinoic acid-induced gene 3 protein |
| Simboli geni | GPRC5C |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?