Learn More
Fisherbrand™ Grade 603 General Purpose Filter Paper
Suitable for all general works in laboratories. Fisherbrand™ Grade 603 General Purpose Filter Paper is thin and has fast filtration velocity.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Applications:- Used in hospitals, research laboratories, schools, chemical industries, pharmaceutical, cosmetic, food industries
Specifica
Sheet | |
35 to 65 μm | |
603 | |
0.14 mm | |
520 mm |
35 to 65μm | |
General works, hospitals, research laboratories, schools, chemical industries, pharmaceutical, cosmetic, food and beverage, etc | |
500 Pack | |
520 x 420mm | |
420 mm |