Learn More
Kartell™ Graduated Conical Test Tubes
Manufactured with high-transparency polypropylene for strength and good visibility. Karell™ Graduated Conical Test Tubes deliver stability and convenience for sample collection and transportation.
Marca: Kartell™ 8400400
Dettagli aggiuntivi : Peso : 6.84000kg
Descrizione
- Self-standing bottom provides flexibility in the lab
- Permanent molded graduations are visible with any color liquid
- Withstands temperatures down to - 80°C
- Supplied with polypropylene tight screw cap for storage and transportation