Learn More
Kartell™ Graduated Conical Test Tubes
Manufactured with high-transparency polypropylene for strength and good visibility. Karell™ Graduated Conical Test Tubes deliver stability and convenience for sample collection and transportation.
Marca:  Kartell™ 8400400
Dettagli aggiuntivi : Peso : 6.84000kg
Codice prodotto. 10584801
  • 204.00€ / pezzi 500
Spedizione stimata: 16-05-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Self-standing bottom provides flexibility in the lab
  • Permanent molded graduations are visible with any color liquid
  • Withstands temperatures down to - 80°C
  • Supplied with polypropylene tight screw cap for storage and transportation