Learn More
Thermo Scientific™ Graphite Ferrule
Product Code. 11583331 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With:
0.1 to 0.32mm GC Column
0.45 to 0.53 mm GC Column
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Unit Size
11583331 0.1 to 0.32mm GC Column Pack of 10
11573331 0.45 to 0.53 mm GC Column Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11583331 Supplier Thermo Scientific™ Supplier No. 290GP132
Spedizione stimata: 02-12-2026
Ensure reliable connections for Thermo Scientific™ gas chromatography instruments with these Thermo Scientific™ 100% Graphite Ferrules for Thermo Scientific Instruments.
  • Engineered for dependable laboratory performance and consistent results
  • Manufactured to meet Thermo Scientific quality standards
  • Designed for compatibility with common laboratory instruments and workflows
  • Helps improve reproducibility and sample integrity
  • Suitable for research, analytical, and routine laboratory applications
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
For Use With 0.1 to 0.32mm GC Column
Material Graphite