Learn More
Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator
Codice prodotto. 15422550
Cambia vista
Click to view available options
Capacità (sistema metrico):
2.5 mL
3.5 mL
8 mL
13 x 100 mm
Colore chiusura:
Red with White Ring
Red with Yellow Ring
Dimensione della confezione:
pezzi 1200
pezzi 50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Colore chiusura Dimensione della confezione
15422550 8 mL Red with Yellow Ring pezzi 50
15422530 3.5 mL Red with Yellow Ring pezzi 50
13008652 3.5 mL Red with Yellow Ring -
10144534 8 mL Red with Yellow Ring -
15484641 13 x 100 mm Red with Yellow Ring -
13068682 2.5 mL Red with White Ring pezzi 1200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
\u00C8 possible ordinare fino a esaurimento dello stock disponibile. Qualora un articolo alternativo sia stato identificato, lo stesso sar\u00E0 visibile qui di seguito
Codice prodotto. 15422550 Fornitore Greiner Bio-One N. del fornitore 455071
  • ONLINE EXCLUSIVE PRICE
    50.40€
    19.56€ / pezzi 50
    Risparmia 30.84€
    61% Sconto
 Articolo soggetto a restrizioni
In stock
\u00C8 possible ordinare fino a esaurimento dello stock disponibile. Qualora un articolo alternativo sia stato identificato, lo stesso sar\u00E0 visibile qui di seguito
For routine clinical chemistry, immunohematology, serology or immunology testing
  • Coated with micronized silica particles which activate clotting when tubes are gently inverted
  • Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Pediatric tubes come with white rings for quick identification
  • Safety Twist Caps can be removed and replaced several times
  • Nonscrew caps can be removed and replaced as often as required
Compliance
Tubes with gel separator are FDA 510(k) cleared for therapeutic drug monitoring (TDM)
TRUSTED_SUSTAINABILITY
Specifica
Additivo Microscopic Silica Particles with Separation Gel
Materiale Plastic
Tipo di chiusura Non-ridged Pull Cap
Capacità (sistema metrico) 8 mL
Colore chiusura Red with Yellow Ring