Learn More
Merck Accessori porta filtri in vetro da 90 mm
Accessories for filter holders for sample preparation.
Marca: Merck XX1014745
584.99 EUR valido fino al 2024-12-31
Usa il codice promo "23269" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.61730kg
Descrizione
These accessory products are designed to replace parts contained in Millipore glass vacuum filtration kits or provide for an alternative configuration.This item is an accessory part for the Millipore All-Glass Filter Holders Kits and Millipore MilliSolve Kit.Specifica
Clear | |
Standard bottom | |
Borosilicate Glass | |
4 L |