missing translation for 'onlineSavingsMsg'
Learn More

GSK1016790A, MedChemExpress

Codice prodotto. 16493767
Encompass_Preferred
Change view
Click to view available options
Quantità:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
16493767 50 mg -
16470278 5 mg 5mg
16457646 100 mg 100mg
16437818 10 mg 10mg
16403666 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16493767 Supplier Medchem Express Supplier No. HY19608

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

MedChemExpress GSK1016790A is a potent and selective transient receptor potential vanilloid 4 (TRPV4) channel agonist. GSK1016790A can elicit Ca2+ influx and elevate intracellular Ca2+ in HEK cells.

GSK1016790A is a potent and selective transient receptor potential vanilloid 4 (TRPV4) channel agonist. GSK1016790A can elicit Ca2+ influx and elevate intracellular Ca2+ in HEK cells.

Specifications

Materiale o nome chimico GSK1016790A
CAS 942206-85-1
Colore Off-White
Note sul grado di purezza Research
Formula molecolare C28H32Cl2N4O6S2
Quantità 50 mg
Informazioni di solubilità DMSO : ≥ 33 mg/mL (50.33 mM)
SMILES O=C(C1=CC2=CC=CC=C2S1)N[C@H](C(N3CCN(C([C@@H](NS(=O)(C4=CC=C(Cl)C=C4Cl)=O)CO)=O)CC3)=O)CC(C)C
Molecular Weight (g/mol) 655.61
Peso formulazione 655.61
Percent Purity 98.09%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.