Learn More
Checkers Guard Dog™ 3 Channel Cable Protector
Product Code. 17742408
Change view
Click to view available options
Color:
Black, Orange
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17742408 Black, Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17742408 Supplier Checkers Supplier No. PGDS0003OB
  • ONLINE EXCLUSIVE PRICE
    477.49€
    387.90€ / pezzi 1
    Risparmia 89.59€
    18% Sconto
Spedizione stimata: 26-11-2026
Guard Dog 3-channel cable protector with hinged lid provides a safer crossing for vehicle and pedestrian traffic while protecting valuable electrical cables from damage. Its modular interlocking design allows you to extend to any length.
  • Guard Dog series 3-channel cable protector with hinged lid provides a safer crossing for vehicle and pedestrian traffic while protecting valuable electrical cables and hose lines from damage
  • Modular interlocking design is ideal for use in amusement parks, entertainment venues, sporting events, and most public, commercial, industrial, construction, and utility applications
Specifications
Includes Hinged Lid
Length (English) 20.1 in.
Length (Metric) 51 cm
Width (Metric) 91 cm
Color Black, Orange
Type Cable Protector
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.