Learn More
Checkers Guard Dog™ 5 Channel Cable Protector
Product Code. 17772408
Change view
Click to view available options
Color:
Black, Orange
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17772408 Black, Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17772408 Supplier Checkers Supplier No. PGDS0007OB
  • ONLINE EXCLUSIVE PRICE
    329.88€
    268.21€ / pezzi 1
    Risparmia 61.67€
    18% Sconto
Spedizione stimata: 26-11-2026
Guard Dog 5-channel cable protector with hinged lid provides a safer crossing for vehicle and pedestrian traffic while protecting valuable electrical cables from damage. Its modular interlocking design allows you to extend to any length.
  • Guard Dog series 5-channel cable protector with hinged lid provides a safer crossing for vehicle and pedestrian traffic while protecting valuable electrical cables and hose lines from damage
  • Modular interlocking design is ideal for use in amusement parks, entertainment venues, sporting events, and most public, commercial, industrial, construction, and utility applications
Specifications
Length (English) 36 in.
Length (Metric) 91 cm
Width (Metric) 50 cm
Color Black, Orange
Material Polyurethane
Type Cable Protector