Learn More
Checkers Guard Dog™ 5 Channel End Caps
Product Code. 17752418
Change view
Click to view available options
Color:
Orange
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
17752418 Orange Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17752418 Supplier Checkers Supplier No. PGDS0019OR
  • ONLINE EXCLUSIVE PRICE
    150.96€
    121.93€ / pezzi 1
    Risparmia 29.03€
    19% Sconto
Spedizione stimata: 26-11-2026
Guard Dog™ 5 Channel End Caps feature a load capacity of 9526 kg/axle.
  • Guard Dog series 5-Channel End Caps provides a safer crossing for vehicle and pedestrian traffic while protecting valuable electrical cables and hose lines from damage
  • Modular interlocking design is ideal for use in amusement parks, entertainment venues, sporting events, and most public, commercial, industrial, construction, and utility applications
Specifications
Dimensions (L x W x H) 50 x 22 x 5 cm
Color Orange
Material Polyurethane
Type End Cap