Learn More
H-151, MedChemExpress
Codice prodotto. 16484678
Encompass_Preferred
Cambia vista
Click to view available options
Quantità:
5 mg
10 mg
50 mg
100 mg
Dimensione della confezione:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
16484678 10 mg 10mg
16480967 50 mg 50mg
16445516 100 mg 100mg
16427397 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 16484678 Fornitore Medchem Express N. del fornitore HY112693
MedChemExpress H-151 is a potent, selective and covalent antagonist of STING that has noteworthy inhibitory activity both in cells and in vivo. H-151 reduces TBK1 phosphorylation and suppresses STING palmitoylation. H-151 can be used for the research of autoinflammatory disease.
H-151 is a potent, selective and covalent antagonist of STING that has noteworthy inhibitory activity both in cells and in vivo. H-151 reduces TBK1 phosphorylation and suppresses STING palmitoylation. H-151 can be used for the research of autoinflammatory disease.
Specifica
Materiale o nome chimico H-151
Forma fisica Powder
CAS 941987-60-6
Colore White
Quantità 10 mg
Note sul grado di purezza Research
Formula molecolare C17H17N3O
Da utilizzare con (applicazione) COVID-19-immunoregulation
Informazioni di solubilità DMSO : 31.25 mg/mL (111.87 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(NC1=CNC2=C1C=CC=C2)NC3=CC=C(CC)C=C3
Molecular Weight (g/mol) 279.34
Peso formulazione 279.34
Percent Purity 97.08%
Grado Research
Research purposes only