Learn More
H2L5186303, R&D Systems™
Codice prodotto. 30001636
Cambia vista
Click to view available options
Quantità:
10 mg
50 mg
Dimensione della confezione:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
30001636 50 mg 50mg
30001621 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 30001636 Fornitore Tocris Bioscience N. del fornitore 4878/50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
A Tocris product, part of R&D Systems. Potent and selective LPA2 receptor antagonist
Potent and selective lysophosphatidic acid 2 (LPA2) receptor antagonist (IC50 values are 8.9, 1230 and 27354 nM for LPA2, LPA3 and LPA1 receptors respectively, in a LPA-elicited calcium mobilization assay).
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 139262-76-3
Formula molecolare C26H20N2O8
Molecular Weight (g/mol) 488.452
InChI Key HZFPOTBCYPWQSH-DZDAAMPGSA-N
Sinonimo z,z-4,4'-1,3-phenylenebis oxy-4,1-phenyleneimino bis 4-oxo-2-butenoic acid, 2z-3-4-3-4-2z-3-carboxyprop-2-enamido phenoxy phenoxy phenyl carbamoyl prop-2-enoic acid
PubChem CID 90488981
IUPAC Name (Z)-4-[4-[3-[4-[[(Z)-3-carboxyprop-2-enoyl]amino]phenoxy]phenoxy]anilino]-4-oxobut-2-enoic acid
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)NC(=O)C=CC(=O)O)OC3=CC=C(C=C3)NC(=O)C=CC(=O)O
Specifications
Materiale o nome chimico H2L5186303
Quantità 50 mg
Peso formulazione 488.45
Percent Purity >98%
Product Line Tocris