Learn More
CleanSpace™ HALO Power System
Product Code. 15900102 Shop All CleanSpace Technology Products
Change view
Click to view available options
Power Unit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15900102 Power Unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15900102 Supplier CleanSpace™ Supplier No. CS3000
  • ONLINE EXCLUSIVE PRICE
    1773.23€
    1046.06€ / pezzi 1
    Risparmia 727.17€
    41% Sconto
View all promotions
 Restricted Item
Spedizione stimata: 23-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Battery-powered, positive pressure air-purifying respirator with IP Rating 66. Suitable for healthcare, laboratory, pharmaceutical and emergency response. Protection in applications with particulate contaminants and biohazards as well as heavy water and decontamination procedures.
  • IP Rating 66 – resistant to dust and liquid ingress
  • Suitable for decontamination procedures
  • Light weight (350g),compact and comfortable
  • Long operating time: up to 9 hours and quick battery recharging: < 2hours
  • AirSensit™ System for mask pressure control and breath responsive airflow
  • One-button system and standby and auto-start/stop mode for quick don/doffing
  • Easy to use support harness
  • Altitude compensated filter blocked detection
  • Easy and accurate flow capability check
Specifications
Product Type PAPR Assembly, No Headtop
Model CS3000
Mask sold separately