Learn More
Hamilton™ Chemically Inert Manual Valves
Codice prodotto. 11562425
Cambia vista
Click to view available options
Tipo:
1-1 Manual Valve
3-2 Manual Valve
3-3 Manual Valve
Dimensione della confezione:
Pezzo
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo Dimensione della confezione
11562425 1-1 Manual Valve Pezzo
10337272 3-3 Manual Valve Pezzo
10217362 3-2 Manual Valve Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 11562425 Fornitore Hamilton™ N. del fornitore 86726
  • ONLINE EXCLUSIVE PRICE
    148.00€
    129.22€ / pezzi 1
    Risparmia 18.78€
    12% Sconto
Spedizione stimata: 15-12-2026
Valves have a fluid path containing PTFE and CTFE
The ports on the valve are 1/4-28 threads and come in different flow paths.
Specifica
Materiale CTFE, PTFE
Tipo 1-1 Manual Valve