Learn More
Hamilton™ PRP-C18 Reversed-Phase HPLC Column Accessories - Guard Cartridges
Guard columns protect HPLC columns by removing particulate contaminants and highly adsorptive compounds from samples thus prolonging analytical column life
264.00€ - 335.00€
Specifica
Dimensione particelle 12 to 20 μm
Pressione max 5000 psi
Diametro (sistema metrico) 1.5875 mm (0.0625 in.)
Visualizza altre specifiche
Prodotti 2
Codice del prodotto Marca Tipo Materiale Prezzo Quantità e disponibilità  
Codice del prodotto Marca Tipo Materiale Prezzo Quantità e disponibilità  
15335074
Vedere i documenti Dettagli della promozione
Hamilton™
79686
Analytical Guard Column Replacement Cartridges Stainless Steel
335.00€
pezzi 5
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 30-04-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
15315074
Vedere i documenti Dettagli della promozione
Hamilton™
79690
Semi-Prep/Prep Guard Column Replacement Cartridges Stainless Steel
264.00€
pezzi 2
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 30-04-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Guard columns are modular and cartridge replacement is easy and tool-free. The desing is optimized to reduce dead volume and prevent extra column band broadening. As with all guard columns these should be installed before the analytical column and after the injection valve.
Guard Cartridges for Stainless Steel HPLC Columns
  • Housing Material: Anodized aluminum
  • Connecting Tubing Size and material: 1/16 in. O.D., 0.28mm I.D., 316 stainless-steel
  • Cartridge Materials: Barrel: CTFE; Sleeve: 304 stainless-steel; Frits: 316 stainless-steel; Frit Porosity: 2μm
  • Cartridge Dimension: 20 x 2.3mm I.D.
  • Cartridge Bed Volume: 80μL
  • Cartridge Packing Material: As labeled on the cartridge
Semi-prep/Preparative Guard Cartridges for Stainless Steel HPLC Columns
  • Housing Material: Anodized aluminum
  • Connecting Tubing Size and material: 1/16 in. O.D., 0.28mm I.D., 316 stainless-steel
  • Cartridge Materials: Barrel: CTFE; Sleeve: 304 stainless-steel; Frits: 316 stainless-steel; Frit Porosity: 2μm
  • Cartridge Dimension: 20 x 4.6mm I.D.
  • Cartridge Bed Volume: 332μL
  • Cartridge Packing Material: As labeled on the cartridge
Guard Cartridges for PEEK HPLC Columns
  • Housing Material: Polyetheretherketone (PEEK) and Delrin
  • Connecting Tubing Size and material: 1/16 in. O.D., 0.25mm I.D., PEEK
  • Cartridge Materials: Barrel: PEEK; Frits: Titanium; Frit Porosity: 2μm
  • Cartridge Dimension: 8.0 x 3.0mm I.D.
  • Cartridge Bed Volume: 56μL
  • Cartridge Packing Material: As labeled on the cartridge
Specifica
Specifica
12 to 20 μm
5000 psi
1.5875 mm (0.0625 in.)