Learn More
Brady™ Ultimate Security Padlock
Product Code. 15643537 Shop All Brady Products
Change view
Click to view available options
Height (Metric) Shackle:
26.00 mm
50.00 mm
75.00 mm
Unit Size:
Each
Pack of 2
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Shackle Unit Size
15643537 50.00 mm Pack of 2
15633537 26.00 mm Pack of 2
11726785 75.00 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15643537 Supplier Brady™ Supplier No. 805838
  • ONLINE EXCLUSIVE PRICE
    264.00€
    238.25€ / pezzi 2
    Risparmia 25.75€
    9% Sconto
Spedizione stimata: 18-12-2026
The 7 pin disk cylinder gives ultimate protection against manipulation and key interchanges.
  • Shackle and body are made of highest standard hardened alloy steel
Specifications
Product Type Ultimate Security Padlock
Key Type Keyed Different
Alternate Part Number SECURITY PADLOCK 55 MM SHACKLE 50 MM 2
Color Black
Material Hardened Alloy Steel
Description Ultimate Security Padlock
Diameter (English) Shackle 0.433 in.
Diameter (Metric) Shackle 11 mm
Dimensions (L x W) 63.00 x 109.00 mm
For Use With (Application) Lockout Tagout
Height (Metric) 109 mm
Height (Metric) Shackle 50.00 mm
Material (Shackle) Hardened Alloy Steel
No. of Pins 7
Quantity 1 Ea.
Width (Metric) 63 mm
Width (Metric) Shackle 49.50 mm