Learn More
Horst™ HBQ Model Very Flexible Heating Tape
Product Code. 10486371
Change view
Click to view available options
Length (Metric):
0.5 m
1 m
1.5 m
2 m
3 m
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10486371 2 m Each
10332322 0.5 m Each
10087141 1 m Each
10322462 1.5 m Each
10148781 3 m Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10486371 Supplier Horst™ Supplier No. 20404
  • ONLINE EXCLUSIVE PRICE
    670.95€
    413.96€ / pezzi 1
    Risparmia 256.99€
    38% Sconto
View all promotions
Spedizione stimata: 09-12-2026
Specifications
Dimensions (L x W) 2 Lm
Dimensions (L x W x H) 2 Lm
Length (Metric) 2 m
Electrical Requirements ∼230V Voltage
Model HBQ
Product Type Very Flexible Heating Tape