missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDNL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA550619
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
The antibody detects endogenous levels of total HCA-4 protein.
NDNL2 Polyclonal antibody specifically detects NDNL2 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifica
NDNL2 | |
Polyclonal | |
Unconjugated | |
NSMCE3 | |
5730494G16Rik; AI642138; BB044375; HCA4; Hepatocellular carcinoma-associated protein 4; hepatocellular carcinoma-associated protein HCA4; Mageg1; mage-g1; MAGE-G1 antigen; MAGEL3; magg1; melanoma antigen, family G, 1; melanoma-associated antigen G1; NDNL2; necdin-like 2; necdin-like 2, SMC5-SMC6 complex component; necdin-like gene 2; necdin-like protein 2; Non-SMC element 3 homolog; non-structural maintenance of chromosomes element 3 homolog; NSE3; NSE3 homolog, SMC5-SMC6 complex component; NSMCE3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56160, 66647 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q96MG7, Q9CPR8 | |
NSMCE3 | |
Synthetic peptide corresponding to residues near the C terminal of human necdin-like 2 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |