Learn More
Uvex™ He Polo Shirt 9872 ESD
Catalog Number p-7232085
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
White cross- striped ESD Polo Shirt
  • ESD Polo Shirt
  • Color: white
  • Material: 54% Cotton, 44% Polyester, 2% Resistat 185 g/m² Cross- striped ESD Polo Shirt According to EN 61340 high wearing comfort.
  • Vorderteil mit Querpass
  • Polo-colar with visible double button bar
  • Certified in accordance to EN 61340-5-1
  • Size: S(44/46) XXL(60/62)
  • Care: washable with 60°C do not chlorinate, 2 point ironing, dry- cleaning: P, drying with reduced heat treatment
Size Small
Sesso Men's
Colore White
Tipo di chiusura Buttons
Monouso No
Spessore 185 g/m²