Descrizione
- Eliminates evaporation in PCR
- Adjustable sealing temperatures for more flexibility
- Seals 96- and 384-well plates
- Sealing time selectable from 1 to 9 seconds in 0.5 second increments
- Integrated thermostat prevents overheating
- Powerful handle for ergonomic operation
- PCR plate adapter included
Specifica
Specifica
| Tensione | 100/230 V |
| Dimensioni (L x P x A) | 220 × 321 × 425 mm |
| Requisiti elettrici | 100/230 V, 50/60 Hz |
| Da utilizzare con (apparecchiatura) | 96- and 384-well plates |
| Tipo di spina | EU Plug |
| Intervallo temperatura sigillatura | 125°C to 200°C in 1°C increments |
| Frequenza | 50/60 Hz |