Learn More
Cole-Parmer™ Heavy-Duty Aluminum Foil
Product Code. 11720109 Shop All Cole-Parmer Products
Change view
Click to view available options
Length (English):
500 ft.
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (English) Unit Size
11720109 500 ft. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11720109 Supplier Cole-Parmer™ Supplier No. 0627505
Save Up to 60% on Microbiology Essentials
Use promo code 28004
View all promotions
Spedizione stimata: 07-01-2027
This Heavy-Duty Aluminum Foil is an all-purpose foil used for freezer storage, wrapping, sealing, autoclaving, lining, and weighing or transferring samples.
  • Packaged in a dispenser box with serrated cutting strip.
Specifications
Applications Freezer storage, wrapping, sealing, autoclaving, lining, and weighing or transferring samples
Length (English) 500 ft.
Product Type Heavy-Duty Aluminum Foil
Width (English) 18 in.
Width (Metric) 45.7 cm
Length (Metric) 152.4 m
Material Aluminum