missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HEBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA567380
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Immunogen sequence: EVAYEERACEG GKFATVEVTD KPVDEALREA MPKVAKYA Highest antigen sequence identity to the following orthologs - mouse 87%, rat 87%.
HEBP1 Polyclonal antibody specifically detects HEBP1 in Human samples. It is validated for Immunocytochemistry.Specifica
HEBP1 | |
Polyclonal | |
Unconjugated | |
HEBP1 | |
HBP; Hebp; Hebp1; heme binding protein 1; heme-binding protein 1; p22 HBP; p22HBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50865 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRV9 | |
HEBP1 | |
Recombinant Human HEBP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only