Learn More
Heidolph™ Hei-VAP Core ML G3B
Codice prodotto. 16267910 Sfoglia Tutto Heidolph Instruments Prodotti
Cambia vista
Click to view available options
Tipo di spina:
EU Plug
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo di spina Dimensione della confezione
16267910 EU Plug Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 16267910 Fornitore Heidolph™ N. del fornitore 5720131000
Equipment and Instrument End of Year Sale
Usa il codice promozionale 28000
Visualizza tutte le promozioni
Spedizione stimata: 10-11-2026
An operating concept that is focused on the essentials: clear and with access to the basic parameters rotation speed and heating bath temperature at any time.
  • Clear digital display showing actual and set values
  • Two separate knobs with dynamic control for setting the rotation speed and heating bath temperature: fast turning for large jumps, slow turning for precise setting of the set values
  • Accidental changing of the values can be prevented by the lock function
  • Intuitive LED ring light system makes activated functions clearly visible even from a distance
  • Easy adjustment of the immersion depth up to 155 mm and the inclination angle from 20° to 80°
  • Standby function and residual heat warning at heating bath temperatures above 50°C
  • Motor lift model with transparent plastic coated G3 vertical glassware
  • The standby button stops all functions and, when using a motor lift model, lifts the evaporating flask from the heating bath
TRUSTED_SUSTAINABILITY
Specifica
Tipo Motor Lift Rotary Evaporator
Tipo di spina EU Plug