Descrizione
- Clear digital display showing actual and set values
- Two separate knobs with dynamic control for setting the rotation speed and heating bath temperature: fast turning for large jumps, slow turning for precise setting of the set values
- Accidental changing of the values can be prevented by the lock function
- Intuitive LED ring light system makes activated functions clearly visible even from a distance
- Easy adjustment of the immersion depth up to 155 mm and the inclination angle from 20° to 80°
- Standby function and residual heat warning at heating bath temperatures above 50°C
- Motor lift model with transparent plastic coated G6 vertical glassware for reflux distillation
- The standby button stops all functions and, when using a motor lift model, lifts the evaporating flask from the heating bath
Specifica
Specifica
| Tipo | Motor Lift Rotary Evaporator |
| Tipo di spina | EU Plug |