Learn More
ACROBiosystems HEK293/Human APP (GFP) Stable Cell Line
Codice prodotto. 31453646 Sfoglia Tutto ACROBiosystems Prodotti
Cambia vista
Click to view available options
Quantità:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
31453646 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 31453646 Fornitore ACROBiosystems N. del fornitore CHEKATP0812Vials
 Articolo soggetto a restrizioni
Spedizione stimata: 21-09-2026
Amyloid precursor protein (APP) is a type I integral membrane protein ubiquitously expressed in many tissues and concentrated in the synapses of neurons.
Amyloid precursor protein (APP) is a type I integral membrane protein ubiquitously expressed in many tissues and concentrated in the synapses of neurons. The majority of APP is cleaved at the plasma membrane by the α-secretase in the non-amyloidogenic pathway. The amyloidogenic pathway starts with β-secretase cleavage by BACE1 on the N-terminal part of the Aβ domain, releasing sAPPβ from a membrane-anchored fragment named βCTF or C103, which is subsequently cleaved by γ-secretase to release Aβ.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 2 Vials