Learn More
ACROBiosystems HEK293/Human TrkB Stable Cell Line
Codice prodotto. 31451815 Sfoglia Tutto ACROBiosystems Prodotti
Cambia vista
Click to view available options
Quantità:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
31451815 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 31451815 Fornitore ACROBiosystems N. del fornitore CHEKATP0822Vials
 Articolo soggetto a restrizioni
Spedizione stimata: 21-09-2026
This gene encodes a member of the neurotrophic tyrosine receptor kinase(NTRK)family.This kinase is a membrane-bound receptor that upon neurotrophin binding,phosphorylates itself and members of the MAPK pathway.Signalling through this kinase leads to cell differentiation.Mutations in this gene have been associated with obesity and mood disorders.
This gene encodes a member of the neurotrophic tyrosine receptor kinase(NTRK)family.This kinase is a membrane-bound receptor that upon neurotrophin binding,phosphorylates itself and members of the MAPK pathway.Signalling through this kinase leads to cell differentiation.Mutations in this gene have been associated with obesity and mood disorders.
TRUSTED_SUSTAINABILITY
Specifica
Quantità 2 Vials