Learn More
Angelantoni Industrie™ Helios Series Horizontal Laminar Flow
Codice prodotto. 11925078
Encompass
Cambia vista
Click to view available options
Modello:
Helio 48C
Helio 72C
Include:
Ventilation system: One fan
Ventilation system: Two fan
Dimensione della confezione:
Pezzo
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Modello Include Dimensione della confezione
11925078 Helio 48C Ventilation system: One fan Pezzo
11935078 Helio 72C Ventilation system: Two fan Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11925078 Fornitore Angelantoni Industrie™ N. del fornitore ST00275400000
 Articolo soggetto a restrizioni
Questo articolo richiede un aggiornamento del prezzo prima di essere ordinato.
Richiedi qui il prezzo per questo articolo.
Used to protect the product from dangerous effects due to uncontrolled diffusion of air contaminants during its handling.
  • Sterility of the working area greater than the ISO class 5 EN 14644 1 of particles 0.3mm and 0.5mm
  • Horizontal laminar flow speed of 0.40m/sec. ±20%
  • Stainless steel worktop AISI 304L 2B finishing
  • Tempered glass side walls
  • Worktop lighting with two fluorescent lamps
  • Fans are of the centrifugal type with IP55 protection
  • Air filtration performed with pre-filters to lose high retention capacity and H14 HEPA filters
  • Microprocessor control system
  • Automatic air speed controller with digital display
  • N°3 through holes on each side glass for any installations of faucets
Specifica
Tipo di filtro H14 Hepa
Tipo di illuminazione Fluorescent
Materiale Stainless Steel (304L) 2B Finish
Include Ventilation system: One fan
Modello Helio 48C