missing translation for 'onlineSavingsMsg'
Learn More

HELLS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11955-1-AP-20UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16899363

  • 129.00€ / 20µL

Spedizione stimata: 23-12-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

This gene encodes a member of the polycystin protein family. The encoded protein contains multiple transmembrane domains, and cytoplasmic N- and C-termini. The protein may be an integral membrane protein involved in cell-cell/matrix interactions. The encoded protein may function in renal tubular development, morphology, and function, and may modulate intracellular calcium homoeostasis and other signal transduction pathways. This protein interacts with polycystin 1 to produce cation-permeable currents. Mutations in this gene have been associated with autosomal domit polycystic kidney disease.
Specifica

Specifica

HELLS
Polyclonal
Unconjugated
HELLS
helicase, lymphoid, HELLS, LSH, Lymphoid specific helicase, Nbla10143, PASG, SMARCA6
Rabbit
Antigen Affinity Chromatography
RUO
15201, 294071, 3070
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q60848, Q9NRZ9
Hells
HELLS Fusion Protein Ag2558
20 μL
Primary
Mouse, Human, Rat
Antibody
IgG
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali